Assay Details
| Gene Symbol: CAPZB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113444.2 | NP_001106915.1 | 3 - 4 | 318 | 160 | |
| XM_005656029.3 | XP_005656086.1 | 354 | 160 | |||
| XM_021093206.1 | XP_020948865.1 | 243 | 160 | |||
| XM_021093208.1 | XP_020948867.1 | 243 | 160 | |||
| GenBank mRNA | AK399057.1 | - | 3 - 4 | 319 | 160 | |
| AY609408.1 | - | 3 - 4 | 258 | 160 | ||
| EF202986.1 | - | 3 - 4 | 320 | 160 | ||
| GU165833.1 | - | 2 - 3 | 216 | 160 | ||
| GU165834.1 | - | 2 - 3 | 216 | 160 | ||
| JX569751.1 | - | 3 - 4 | 316 | 160 | ||
| JX569752.1 | - | 3 - 4 | 316 | 160 | ||
| JX966416.1 | - | 3 - 4 | 316 | 160 | ||
More Information
Set Membership: |
|

CAPZB
Genomic Map