Assay Details
| Gene Symbol: KLF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134349.2 | NP_001127821.1 | 1 - 2 | 165 | 123 | |
| GenBank mRNA | AK233450.1 | - | 1 - 2 | 175 | 123 | |
| EF095723.2 | - | 1 - 2 | 165 | 123 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001134349.2 transcripts.
Set Membership: |
|

KLF15
Genomic Map