Assay Details
| Gene Symbol: BCO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136512.1 | NP_001129984.1 | 4 - 5 | 620 | 61 | |
| XM_021093242.1 | XP_020948901.1 | 2091 | 61 | |||
| XM_021093243.1 | XP_020948902.1 | 828 | 61 | |||
| XM_021093244.1 | XP_020948903.1 | 738 | 61 | |||
| XM_021093245.1 | XP_020948904.1 | 1062 | 61 | |||
| XM_021093246.1 | XP_020948905.1 | 1800 | 61 | |||
| XM_021093247.1 | XP_020948906.1 | 495 | 61 | |||
| XM_021093248.1 | XP_020948907.1 | 631 | 61 | |||
| GenBank mRNA | FJ211199.1 | - | 4 - 5 | 620 | 61 | |
More Information
Set Membership: |
|

BCO1
Genomic Map