Assay Details
| Gene Symbol: RAD18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142833.1 | NP_001136305.1 | 4 - 5 | 345 | 73 | |
| XM_021068321.1 | XP_020923980.1 | 618 | 73 | |||
| XM_021068322.1 | XP_020923981.1 | 616 | 73 | |||
| XM_021068323.1 | XP_020923982.1 | 627 | 73 | |||
| GenBank mRNA | AK351874.1 | - | 5 - 6 | 477 | 73 | |
| FJ236699.1 | - | 4 - 5 | 345 | 73 | ||
More Information
Set Membership: |
|

RAD18
Genomic Map