Assay Details
| Gene Symbol: GDF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174056.1 | NP_001167527.1 | 1 - 2 | 333 | 95 | |
| GenBank mRNA | GU386349.1 | - | 1 - 2 | 333 | 95 | |
More Information
Set Membership: |
|

GDF15
Genomic Map