Assay Details
| Gene Symbol: GLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177925.1 | NP_001171396.1 | 5 - 6 | 815 | 102 | |
| GenBank mRNA | AK234770.1 | - | 5 - 6 | 922 | 102 | |
| AK236208.1 | - | 5 - 6 | 872 | 102 | ||
| AK397923.1 | - | 5 - 6 | 1112 | 102 | ||
| EU561635.1 | - | 5 - 6 | 815 | 102 | ||
More Information
Set Membership: |
|

GLA
Genomic Map