Assay Details
| Gene Symbol: FZD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195339.1 | NP_001182268.1 | 1 | 1522 | 74 | |
| GenBank mRNA | GU373716.1 | - | 1 | 1522 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: FZD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195339.1 | NP_001182268.1 | 1 | 1522 | 74 | |
| GenBank mRNA | GU373716.1 | - | 1 | 1522 | 74 | |
Set Membership: |
|