Assay Details
| Gene Symbol: VARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195378.1 | NP_001182307.1 | 27 - 28 | 3603 | 63 | |
| GenBank mRNA | AK236130.1 | - | 8 - 9 | 975 | 63 | |
| AK397216.1 | - | 9 - 10 | 1052 | 63 | ||
| AK400536.1 | - | 16 - 17 | 1755 | 63 | ||
| EU282356.1 | - | 27 - 28 | 3603 | 63 | ||
More Information
Set Membership: |
|

VARS1
Genomic Map