Assay Details
| Gene Symbol: VARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195378.1 | NP_001182307.1 | 28 - 29 | 3933 | 72 | |
| GenBank mRNA | AK236130.1 | - | 9 - 10 | 1305 | 72 | |
| AK397216.1 | - | 10 - 11 | 1382 | 72 | ||
| AK400536.1 | - | 17 - 18 | 2085 | 72 | ||
| EU282356.1 | - | 28 - 29 | 3933 | 72 | ||
More Information
Set Membership: |
|

VARS1
Genomic Map