Assay Details
| Gene Symbol: RFXANK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243347.1 | NP_001230276.1 | 6 - 7 | 781 | 69 | |
| XM_021080054.1 | XP_020935713.1 | 1060 | 69 | |||
| GenBank mRNA | AK347490.1 | - | 6 - 7 | 784 | 69 | |
| BI404562.1 | - | 3 - 4 | 381 | 69 | ||
| DY426545.1 | - | 5 - 6 | 464 | 69 | ||
More Information
Set Membership: |
|

RFXANK
Genomic Map