Assay Details
| Gene Symbol: TMEM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243567.1 | NP_001230496.1 | 1 - 2 | 150 | 81 | |
| GenBank mRNA | DN112483.1 | - | 1 - 2 | 150 | 81 | |
More Information
Set Membership: |
|

TMEM17
Genomic Map