Assay Details
| Gene Symbol: VAMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243593.1 | NP_001230522.1 | 4 - 5 | 396 | 85 | |
| XM_021079480.1 | XP_020935139.1 | 361 | 85 | |||
| GenBank mRNA | AK230991.1 | - | 3 - 4 | 326 | 85 | |
| AK343727.1 | - | 4 - 5 | 396 | 85 | ||
More Information
Set Membership: |
|

VAMP7
Genomic Map