Assay Details
| Gene Symbol: RPL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243603.1 | NP_001230532.1 | 1 - 2 | 45 | 73 | |
| GenBank mRNA | AK231514.1 | - | 1 - 2 | 46 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243603.1 transcripts.
Set Membership: |
|

RPL37
Genomic Map