Assay Details
| Gene Symbol: RPL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243603.1 | NP_001230532.1 | 3 - 4 | 265 | 85 | |
| GenBank mRNA | AK231514.1 | - | 3 - 4 | 266 | 85 | |
| AK234186.1 | - | 3 - 4 | 256 | 85 | ||
| AK399835.1 | - | 3 - 4 | 265 | 85 | ||
| AY610371.1 | - | 2 - 3 | 232 | 85 | ||
More Information
Set Membership: |
|

RPL37
Genomic Map