Assay Details
| Gene Symbol: DYNC1LI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243620.1 | NP_001230549.1 | 11 - 12 | 1287 | 56 | |
| XM_021093420.1 | XP_020949079.1 | 1320 | 56 | |||
| GenBank mRNA | AK350541.1 | - | 11 - 12 | 1287 | 56 | |
More Information
Set Membership: |
|

DYNC1LI2
Genomic Map