Assay Details
| Gene Symbol: CES3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243625.1 | NP_001230554.1 | 11 - 12 | 1486 | 94 | |
| GenBank mRNA | AK232431.1 | - | 11 - 12 | 1486 | 94 | |
| AK393090.1 | - | 11 - 12 | 1488 | 94 | ||
More Information
Set Membership: |
|

CES3
Genomic Map