Assay Details
| Gene Symbol: CYP19A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214430.1 | NP_999595.1 | 2 - 3 | 175 | 125 | |
| XM_021083457.1 | XP_020939116.1 | 253 | 125 | |||
| GenBank mRNA | U52142.1 | - | 2 - 3 | 175 | 125 | |
| U92245.1 | - | 1 - 2 | 149 | 125 | ||
More Information
Set Membership: |
|

CYP19A2
Genomic Map