Assay Details
| Gene Symbol: GRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB158409.1 | - | 851 | 65 | ||
| AB161682.1 | - | 851 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRP| Gene Symbol: GRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB158409.1 | - | 851 | 65 | ||
| AB161682.1 | - | 851 | 65 | |||
Set Membership: |
|