Assay Details
| Gene Symbol: ETT1-alpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY376128.1 | - | 1154 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ETT1-alpha| Gene Symbol: ETT1-alpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY376128.1 | - | 1154 | 103 | ||
Set Membership: |
|