Assay Details
| Gene Symbol: la-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | M28704.1 | - | 551 | 65 | ||
| X56781.1 | - | 551 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

la-17| Gene Symbol: la-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | M28704.1 | - | 551 | 65 | ||
| X56781.1 | - | 551 | 65 | |||
Set Membership: |
|