Assay Details
| Gene Symbol: Hmgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB272226.1 | - | 428 | 90 | ||
| Z11540.1 | - | 451 | 90 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmgb3| Gene Symbol: Hmgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB272226.1 | - | 428 | 90 | ||
| Z11540.1 | - | 451 | 90 | |||
Set Membership: |
|