Assay Details
| Gene Symbol: RBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ103754.1 | - | 550 | 77 | ||
| EU018460.1 | - | 679 | 77 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBP| Gene Symbol: RBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ103754.1 | - | 550 | 77 | ||
| EU018460.1 | - | 679 | 77 | |||
Set Membership: |
|