Assay Details
| Gene Symbol: LOC100136977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EU293127.1 | - | 354 | 79 | ||
| KJ551526.1 | - | 385 | 79 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100136977| Gene Symbol: LOC100136977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EU293127.1 | - | 354 | 79 | ||
| KJ551526.1 | - | 385 | 79 | |||
Set Membership: |
|