Assay Details
| Gene Symbol: SNAP-ALPHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB001375.1 | - | 6 - 7 | 659 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNAP-ALPHA| Gene Symbol: SNAP-ALPHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB001375.1 | - | 6 - 7 | 659 | 101 | |
Set Membership: |
|