Assay Details
| Gene Symbol: SNAP-ALPHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB001375.1 | - | 8 - 9 | 794 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNAP-ALPHA| Gene Symbol: SNAP-ALPHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB001375.1 | - | 8 - 9 | 794 | 93 | |
Set Membership: |
|