Assay Details
| Gene Symbol: P5CS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ005686.1 | - | 1 - 2 | 117 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

P5CS| Gene Symbol: P5CS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ005686.1 | - | 1 - 2 | 117 | 88 | |
Set Membership: |
|