Assay Details
| Gene Symbol: AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY538746.1 | - | 1 - 2 | 347 | 96 | |
| GU133634.1 | - | 1 - 2 | 181 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AP1| Gene Symbol: AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY538746.1 | - | 1 - 2 | 347 | 96 | |
| GU133634.1 | - | 1 - 2 | 181 | 96 | ||
Set Membership: |
|