Assay Details
| Gene Symbol: FT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002270372.1 | XP_002270408.1 | 2 - 3 | 323 | 73 | |
| GenBank mRNA | DQ504308.1 | - | 2 - 3 | 329 | 73 | |
| DQ871590.1 | - | 2 - 3 | 294 | 73 | ||
| EF157728.1 | - | 2 - 3 | 302 | 73 | ||
More Information
Set Membership: |
|

FT
Genomic Map