Assay Details
| Gene Symbol: AP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EF418603.1 | - | 4 - 5 | 423 | 100 | |
| GU133632.1 | - | 4 - 5 | 423 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AP3| Gene Symbol: AP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EF418603.1 | - | 4 - 5 | 423 | 100 | |
| GU133632.1 | - | 4 - 5 | 423 | 100 | ||
Set Membership: |
|