Assay Details
| Gene Symbol: MLO15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EU360796.1 | - | 10 - 11 | 1168 | 70 | |
| EU812236.1 | - | 10 - 11 | 1168 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLO15| Gene Symbol: MLO15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EU360796.1 | - | 10 - 11 | 1168 | 70 | |
| EU812236.1 | - | 10 - 11 | 1168 | 70 | ||
Set Membership: |
|