Assay Details
| Gene Symbol: pdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001197.1 | NP_001001197.1 | 7 - 8 | 759 | 95 | |
| NM_001017072.2 | NP_001017072.1 | 7 - 8 | 810 | 95 | ||
| XM_012957026.2 | XP_012812480.1 | 8 - 9 | 838 | 95 | ||
| XM_012957027.2 | XP_012812481.1 | 8 - 9 | 956 | 95 | ||
| GenBank mRNA | BC067306.1 | - | 7 - 8 | 759 | 95 | |
| CR761539.2 | - | 7 - 8 | 810 | 95 | ||
More Information
Set Membership: |
|

pdha1
Genomic Map