Assay Details
| Gene Symbol: mindy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001204.1 | NP_001001204.1 | 4 - 5 | 485 | 66 | |
| XM_018094601.1 | XP_017950090.1 | 4 - 5 | 485 | 66 | ||
| GenBank mRNA | BC067311.1 | - | 4 - 5 | 485 | 66 | |
More Information
Set Membership: |
|

mindy3
Genomic Map