Assay Details
| Gene Symbol: olah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001207.1 | NP_001001207.1 | 5 - 6 | 614 | 77 | |
| XM_012964486.2 | XP_012819940.1 | 5 - 6 | 1017 | 77 | ||
| XM_018094603.1 | XP_017950092.1 | 5 - 6 | 621 | 77 | ||
| GenBank mRNA | BC067998.1 | - | 5 - 6 | 614 | 77 | |
More Information
Set Membership: |
|

olah
Genomic Map