Assay Details
| Gene Symbol: ap2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001209.2 | NP_001001209.1 | 7 - 8 | 925 | 61 | |
| XM_012960782.2 | XP_012816236.1 | 7 - 8 | 1101 | 61 | ||
| XM_012960783.2 | XP_012816237.1 | 7 - 8 | 1101 | 61 | ||
| XM_012960784.2 | XP_012816238.1 | 7 - 8 | 1100 | 61 | ||
| GenBank mRNA | BC067918.2 | - | 7 - 8 | 925 | 61 | |
| CR848593.2 | - | 7 - 8 | 902 | 61 | ||
More Information
Set Membership: |
|

ap2a2
Genomic Map