Assay Details
| Gene Symbol: rela | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001211.1 | NP_001001211.1 | 6 - 7 | 742 | 63 | |
| XM_012960785.2 | XP_012816239.1 | 6 - 7 | 783 | 63 | ||
| XM_012960786.2 | XP_012816240.1 | 6 - 7 | 786 | 63 | ||
| GenBank mRNA | BC067968.1 | - | 6 - 7 | 742 | 63 | |
More Information
Set Membership: |
|

rela
Genomic Map