Assay Details
| Gene Symbol: pabpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001230.1 | NP_001001230.1 | 1 - 2 | 396 | 84 | |
| GenBank mRNA | BC067958.1 | - | 1 - 2 | 396 | 84 | |
| CR761856.2 | - | 1 - 2 | 329 | 84 | ||
More Information
Set Membership: |
|

pabpn1
Genomic Map