Assay Details
| Gene Symbol: pabpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001230.1 | NP_001001230.1 | 6 - 7 | 921 | 86 | |
| GenBank mRNA | BC067958.1 | - | 6 - 7 | 921 | 86 | |
| CR761856.2 | - | 6 - 7 | 854 | 86 | ||
More Information
Set Membership: |
|

pabpn1
Genomic Map