Assay Details
| Gene Symbol: rnf139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001242.2 | NP_001001242.1 | 1 - 2 | 230 | 78 | |
| GenBank mRNA | BC067995.2 | - | 1 - 2 | 230 | 78 | |
More Information
Set Membership: |
|

rnf139
Genomic Map