Assay Details
| Gene Symbol: calr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001253.1 | NP_001001253.1 | 1 - 2 | 126 | 92 | |
| GenBank mRNA | BC067917.1 | - | 1 - 2 | 126 | 92 | |
More Information
Set Membership: |
|

calr
Genomic Map