Assay Details
| Gene Symbol: atp5f1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001256.1 | NP_001001256.1 | 4 - 5 | 607 | 93 | |
| GenBank mRNA | BC067388.1 | - | 4 - 5 | 607 | 93 | |
| CR762341.2 | - | 4 - 5 | 659 | 93 | ||
More Information
Set Membership: |
|

atp5f1b
Genomic Map