Assay Details
| Gene Symbol: rhcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003661.1 | NP_001003661.1 | 7 - 8 | 897 | 73 | |
| XM_012958791.2 | XP_012814245.1 | 7 - 8 | 959 | 73 | ||
| XM_012958792.2 | XP_012814246.1 | 6 - 7 | 700 | 73 | ||
| GenBank mRNA | AY271818.1 | - | 7 - 8 | 897 | 73 | |
More Information
Set Membership: |
|

rhcg
Genomic Map