Assay Details
| Gene Symbol: pofut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004774.1 | NP_001004774.1 | 4 - 5 | 766 | 65 | |
| GenBank mRNA | AJ781498.1 | - | 4 - 5 | 766 | 65 | |
| BC082519.1 | - | 4 - 5 | 713 | 65 | ||
More Information
Set Membership: |
|

pofut1
Genomic Map