Assay Details
| Gene Symbol: dlx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004778.1 | NP_001004778.1 | 1 - 2 | 512 | 72 | |
| GenBank mRNA | BC074511.1 | - | 1 - 2 | 512 | 72 | |
| CR760090.2 | - | 1 - 2 | 425 | 72 | ||
More Information
Set Membership: |
|

dlx5
Genomic Map