Assay Details
| Gene Symbol: stk35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004779.1 | NP_001004779.1 | 2 - 3 | 862 | 77 | |
| GenBank mRNA | BC074513.1 | - | 2 - 3 | 862 | 77 | |
| CR761751.2 | - | 2 - 3 | 834 | 77 | ||
More Information
Set Membership: |
|

stk35
Genomic Map