Assay Details
| Gene Symbol: snrpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004780.1 | NP_001004780.1 | 2 - 3 | 533 | 68 | |
| XM_012964505.2 | XP_012819959.1 | 2 - 3 | 534 | 68 | ||
| GenBank mRNA | BC074514.1 | - | 2 - 3 | 533 | 68 | |
| CT025480.2 | - | 2 - 3 | 117 | 68 | ||
More Information
Set Membership: |
|

snrpd1
Genomic Map