Assay Details
| Gene Symbol: mcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004788.1 | NP_001004788.1 | 608 | 71 | ||
| GenBank mRNA | BC074523.1 | - | 608 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mcat| Gene Symbol: mcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004788.1 | NP_001004788.1 | 608 | 71 | ||
| GenBank mRNA | BC074523.1 | - | 608 | 71 | ||
Set Membership: |
|