Assay Details
| Gene Symbol: dnaja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004807.1 | NP_001004807.1 | 1 - 2 | 147 | 62 | |
| GenBank mRNA | BC074569.1 | - | 1 - 2 | 147 | 62 | |
| CT030566.1 | - | 1 - 2 | 86 | 62 | ||
More Information
Set Membership: |
|

dnaja2
Genomic Map