Assay Details
| Gene Symbol: med31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004826.2 | NP_001004826.2 | 2 - 3 | 167 | 99 | |
| GenBank mRNA | BC074608.1 | - | 2 - 3 | 167 | 99 | |
| CR762011.2 | - | 2 - 3 | 124 | 99 | ||
More Information
Set Membership: |
|

med31
Genomic Map