Assay Details
| Gene Symbol: med31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004826.2 | NP_001004826.2 | 3 - 4 | 274 | 83 | |
| NM_001244765.1 | NP_001231694.1 | 2 - 3 | 199 | 83 | ||
| GenBank mRNA | AL887426.2 | - | 2 - 3 | 198 | 83 | |
| BC074608.1 | - | 3 - 4 | 274 | 83 | ||
| CR762011.2 | - | 3 - 4 | 231 | 83 | ||
More Information
Set Membership: |
|

med31
Genomic Map