Assay Details
| Gene Symbol: id4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004839.1 | NP_001004839.1 | 2 - 3 | 781 | 66 | |
| GenBank mRNA | BC074645.1 | - | 2 - 3 | 781 | 66 | |
| CR760882.2 | - | 2 - 3 | 734 | 66 | ||
More Information
Set Membership: |
|

id4
Genomic Map